Order Entry
Canada
Orders LinkContactUsLinkComponent
MacConkey Agar/MacConkey Agar

MacConkey Agar/MacConkey Agar

MacConkey Agar/MacConkey Agar biplate, 15×100 mm biplate, 10 per package
Catalog # 76411-520
Supplier:   Hardy Diagnostics
Supplier #:  J118
MacConkey Agar/MacConkey Agar
Catalog # 76411-520
Supplier:  Hardy Diagnostics
CAS Number:  
MacConkey Agar/MacConkey Agar
MacConkey Agar/MacConkey Agar

MacConkey Agar/MacConkey Agar biplate, 15×100 mm biplate, 10 per package

CAS Number:  
Price
$113.00
Log in for availability
Pack of 10
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

MacConkey Agar for the culture and isolation of gram-negative bacteria, controls swarming of Proteus.

Specifications

Specifications

  • Pack type:
    10 per package
  • Description:
    MacConkey Agar/MacConkey Agar biplate
  • Format:
    15×100 mm biplate

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.