ContactUsLinkComponent
Sanicult™ Hygiene Monitoring Test Kit, Diluent Solution, Sterile
  76423-510
Sanicult™ Hygiene Monitoring Test Kit, Diluent Solution, Sterile
  76423-510
 :  B3OUS5
  • Description:
    5 ml Sanicult sterile diluent solution media fill
  • Environmentally Preferable:
  • Cat. no.:
    76423-510
Swab test kits are for hard-to-reach places, surfaces, viscous liquids, semi-solids, and powders.
They are ideal for applications in food and beverage, engineering, and petrochemical industries. The kit consists of a sterile swab and buffered solution. To use, remove swab from diluent vial, swab sample area, return swab to vial, and mix well. The sample is easily stored and readily transported.
 : 100 per package.