About this item
Transparent cryogenic labels designed to quickly and directly adhere to frozen tubes and cryo vials. Their clear design provides a 'printed-on' effect without concealing the surface or contents underneath. They can be applied under deep-freeze conditions (–80 °C) and withstand prolonged storage in cryogenic conditions (–196 °C), as well as elevated temperatures.
- Temperature range: –196 to 100 °C (–321 to +212 °F)
- Labels can adhere quickly to already frozen vials (no thawing required)
- Ideal for long-term storage in cryogenic conditions (–196 °C)
- Printable with all major thermal-transfer printer brands
These labels are transparent cryogenic labels designed to quickly and directly adhere to frozen tubes and cryo vials. Perfectly suited for long-term cryo-preservation in liquid nitrogen (–196 °C/–321 °F), laboratory freezers (–80 °C/–112 °F), and transportation on dry ice. These versatile clear labels can also withstand temperatures up to 100 °C (212 °F). The labels are printable with all major thermal transfer barcode printer brands or can be inscribed with our permanent ink cryo markers. An overlap of at least 0.25" is recommended to ensure proper adherence to the frozen surface.