Orders LinkContactUsLinkComponent
Cryogenic Piggyback Multi-Layer Barcode Labels
Cryogenic Piggyback Multi-Layer Barcode Labels
Cryogenic Piggyback Multi-Layer Barcode Labels
Certificates
Cryogenic Piggyback Multi-Layer Barcode Labels
Cryogenic Piggyback Multi-Layer Barcode Labels
2 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Designed for long-term cryo-preservation under cryogenic laboratory conditions, these labels allow multiple related labels to be printed at once and placed in one location until the smaller labels within are needed to identify subsequent items.
  • Temperature range: −196 to +110 °C (−321 to +230 °F)
  • Resistant to extreme cryogenic conditions, as low as −196 °C
  • Multi-layer piggyback design facilitates product tracking
  • Labels are resistant to spraying and swabbing with alcohol and surface sanitizing wipes
Cryogenic piggyback labels for identifying small diameter vials and tubes, plates, freezer boxes, and plastic bags. The labels have a label-on-label multi-layer design that allows multiple related labels to be printed at once and placed in one location until the smaller labels within are needed to identify subsequent items. Designed for long-term cryo-preservation under cryogenic laboratory conditions, including liquid and vapor phase liquid nitrogen (−196 °C/−321 °F), −80 °C (−112 °F) freezers, and dry ice transportation. Labels adhere to wet surfaces and are alcohol-resistant. Printable with all major brands of thermal-transfer printers for smudge-proof, serialized alphanumerical information and barcodes.
Product Family Options
Product Information
  • Color
    Core size
    Dimensions
    Packaging
    Availability
    Price
  • 76210-650
    Cryogenic piggyback multi-layer labels
    Cryogenic piggyback multi-layer labels
    White
    1"
    40×40 mm (1.575×1.575")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    Roll of 1,000
    Roll of 1,000 - $380.71
    $380.71
    Roll of 1,000
       $380.71
    Supplier Number: 807656077758
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 76210-652
    Cryogenic piggyback multi-layer labels
    Cryogenic piggyback multi-layer labels
    White
    3"
    40×40 mm (1.575×1.575")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    Roll of 2,000
    Roll of 2,000 - $761.25
    $761.25
    Roll of 2,000
       $761.25
    Supplier Number: 807656077765
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.