Order Entry
Canada
ContactUsLinkComponent
myo-Inositol ≥99% (by HPLC) for microbiology, Millipore®
myo-Inositol ≥99% (by HPLC) for microbiology, Millipore®
Catalog # CA1.04728.0100
Supplier:  MilliporeSigma
CAS Number:  
myo-Inositol ≥99% (by HPLC) for microbiology, Millipore®
Catalog # CA1.04728.0100
Supplier:  MilliporeSigma
Supplier Number:  1.04728.0100
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    100 g
  • Cat. no.:
    CA1.04728.0100
  • Supplier No.:
    1.04728.0100

Specifications

About this item

 :
 :180.16 g/mol
Melting Pt:225-227 °C
Storage Temperature:Ambient
CAS Number:87-89-8
Formula:C₆H₁₂O₆
MDL Number:MFCD00077932
Melting Pt:225…227 °C