Centrifuge Tubes with Conical Base
77587-928
New Product
- Cap included:Yes
- Cap Material:HDPE
- Cap Type:Screw cap (assembled)
- Ø:28 mm
- Sterility:Sterile
- Volume:50 ml
- Color:Transparent
- Length:114 mm
- Material:Polypropylene
- Cat. no.:77587-928
- Packaging:BAGA
- Tube Bottom Shape:Conical base
- Maximum RCF:20000×g
- Cap Color:Natural
- Graduations:Printed graduation (white/blue)
High-quality, safe and robust containers are essential for mixing, processing and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, Sarstedt offers the right product for every laboratory and analytical requirement.
- Base shape: Conical base
- Blue graduated scale for ideal volume reading
- Corresponds to IATA and ADR requirements
- 'Large writing block' allows clear labelling
- Certified free of DNA, DNase, RNase, PCR inhibitors, pyrogens and non-cytotoxic
- Especially leak tight as per the ADR transport directive
- Volume: 50 ml
- Centrifugability up to: 20000×g
- Material: PP
This broad product portfolio in the field of reagent and centrifuge tubes includes options with round, conical or flat base; with or without skirted base; with stopper or screw cap; with or without print or labels.