IOTest 3 Fixative Solution, 100 to 200 Tests, RUO
Catalog # CA77575-366
Supplier: Beckman Coulter
New Product
CAS Number:
Specifications
- Description:IOTest 3 fixative solution, RUO
- Cat. no.:CA77575-366
- No. of Reactions:100 - 200 tests
Specifications
About this item
The IOTest3 Fixative solution allows whole blood or bone marrow specimen preparation by fixing the cell suspension after erythrolysis during immunophenotyping procedures. At least one wash is required between the lysis and the fixation steps.
The IOTest3 Fixative Solution is optimized to work in combination with the IOTest3 Lysing Solution. IOTest Fixative Solution is a formaldehyde-based fixative solution designed for flow cytometry. This reagent is provided as a concentrated solution in PBS.