Orders LinkContactUsLinkComponent
Microbiopsy PathFlow PiSmart Laser Cassettes, Threaded
Supplier:   StatLab
Microbiopsy PathFlow PiSmart Laser Cassettes, Threaded
Supplier:  StatLab
Certificates
Microbiopsy PathFlow PiSmart Laser Cassettes, Threaded
9 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Pathflow PiSmart Laser Cassettes are a two piece design with a rear hinge and front tab closure.
Available in slotted, Biopsy and Microbiopsy. Threaded cassettes come in stacks of 40 cassettes, and 50 stacks per case of 2000. These threaded cassettes can be easily and quickly loaded in the PiSmart Laser Printers. These cassettes are a validated consumable for the PiSmart Laser Cassette Printer.
Product Family Options
Product Information
  • Color
    Availability
    Price
  • 77575-564
    Microbiopsy cassettes with separate lids, with rear hinge and front closure
    Microbiopsy cassettes with separate lids, with rear hinge and front closure
    Blue
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,467.71
    $1,467.71
    Case of 2,000
       $1,467.71
    Supplier Number: CAS-LPI100MBX-BL
  • 77575-566
    Microbiopsy cassettes with separate lids, with rear hinge and front closure
    Microbiopsy cassettes with separate lids, with rear hinge and front closure
    Green
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,467.71
    $1,467.71
    Case of 2,000
       $1,467.71
    Supplier Number: CAS-LPI100MBX-GR
  • 77575-568
    Microbiopsy cassettes with separate lids, with rear hinge and front closure
    Microbiopsy cassettes with separate lids, with rear hinge and front closure
    Gray
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,467.71
    $1,467.71
    Case of 2,000
       $1,467.71
    Supplier Number: CAS-LPI100MBX-GY
  • 77575-570
    Microbiopsy cassettes with separate lids, with rear hinge and front closure
    Microbiopsy cassettes with separate lids, with rear hinge and front closure
    Lavender
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,467.71
    $1,467.71
    Case of 2,000
       $1,467.71
    Supplier Number: CAS-LPI100MBX-LV
  • 77575-572
    Microbiopsy cassettes with separate lids, with rear hinge and front closure
    Microbiopsy cassettes with separate lids, with rear hinge and front closure
    Orange
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,467.71
    $1,467.71
    Case of 2,000
       $1,467.71
    Supplier Number: CAS-LPI100MBX-OR
  • 77575-574
    Microbiopsy cassettes with separate lids, with rear hinge and front closure
    Microbiopsy cassettes with separate lids, with rear hinge and front closure
    Pink
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,467.71
    $1,467.71
    Case of 2,000
       $1,467.71
    Supplier Number: CAS-LPI100MBX-PK
  • 77575-576
    Microbiopsy cassettes with separate lids, with rear hinge and front closure
    Microbiopsy cassettes with separate lids, with rear hinge and front closure
    Tan
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,467.71
    $1,467.71
    Case of 2,000
       $1,467.71
    Supplier Number: CAS-LPI100MBX-TN
  • 77575-578
    Microbiopsy cassettes with separate lids, with rear hinge and front closure
    Microbiopsy cassettes with separate lids, with rear hinge and front closure
    White
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,467.71
    $1,467.71
    Case of 2,000
       $1,467.71
    Supplier Number: CAS-LPI100MBX-WH
  • 77575-580
    Microbiopsy cassettes with separate lids, with rear hinge and front closure
    Microbiopsy cassettes with separate lids, with rear hinge and front closure
    Yellow
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,467.71
    $1,467.71
    Case of 2,000
       $1,467.71
    Supplier Number: CAS-LPI100MBX-YL
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.