Orders LinkContactUsLinkComponent
PathFlow PiSmart Laser Cassettes, Threaded
PathFlow PiSmart Laser Cassettes, Threaded
Supplier:   StatLab
PathFlow PiSmart Laser Cassettes, Threaded
Supplier:  StatLab
Certificates
PathFlow PiSmart Laser Cassettes, Threaded
9 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Pathflow PiSmart laser cassettes are a two piece design with a rear hinge and front tab closure. Threaded cassettes come in stacks of 40 cassettes, and 50 stacks per case of 2000.
These threaded cassettes can be easily and quickly loaded in the PiSmart Laser Printers. These cassettes are a validated consumable for the PiSmart laser cassette printer.
Product Family Options
Product Information
  • Color
    Availability
    Price
  • 77575-546
    PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    Blue
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,419.00
    $1,419.00
    Case of 2,000
       $1,419.00
    Supplier Number: CAS-LPI100BX-BL
  • 77575-550
    PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    Gray
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,419.00
    $1,419.00
    Case of 2,000
       $1,419.00
    Supplier Number: CAS-LPI100BX-GY
  • 77575-548
    PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    Green
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,419.00
    $1,419.00
    Case of 2,000
       $1,419.00
    Supplier Number: CAS-LPI100BX-GR
  • 77575-552
    PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    Lavender
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,419.00
    $1,419.00
    Case of 2,000
       $1,419.00
    Supplier Number: CAS-LPI100BX-LV
  • 77575-554
    PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    Orange
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,419.00
    $1,419.00
    Case of 2,000
       $1,419.00
    Supplier Number: CAS-LPI100BX-OR
  • 77575-556
    PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    Pink
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,419.00
    $1,419.00
    Case of 2,000
       $1,419.00
    Supplier Number: CAS-LPI100BX-PK
  • 77575-558
    PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    Tan
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,419.00
    $1,419.00
    Case of 2,000
       $1,419.00
    Supplier Number: CAS-LPI100BX-TN
  • 77575-560
    PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    White
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,419.00
    $1,419.00
    Case of 2,000
       $1,419.00
    Supplier Number: CAS-LPI100BX-WH
  • 77575-562
    PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
    Yellow
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $1,419.00
    $1,419.00
    Case of 2,000
       $1,419.00
    Supplier Number: CAS-LPI100BX-YL
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)