NANOCOLOR® Photometric Tube Tests, DEHA
Catalog # 77578-490
Supplier: Macherey-Nagel
New Product
CAS Number:
Specifications
- Chemical method:Redox reaction
- Description:Diethylhydroxylamine (DEHA)
- Cat. no.:77578-490
- Range:0.05 - 1.00 mg/L DEHA
- No. of Tests:20
Specifications
About this item
A range of pre-dosed NANOCOLOR® tube tests for photometric determination of DEHA (Bis(2-ethylhexyl)-hexanedioate) in water.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in ext.Ø 16 mm tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.