ContactUsLinkComponent
Optical Bench Kit
Optical Bench Kit
  470356-982
New Product
undefined
Optical Bench Kit
  470356-982
 :  OPTBN-F-6
New Product
  • Description:
    Optical bench kit
  • Environmentally Preferable:
  • Cat. no.:
    470356-982
  • Includes:
    120 cm sturdy metal, graduated, bench, F-LED, object screen, concave and convex lens with 100 mm focal length, concave and convex lens with 200 mm focal length, 6 mounting brackets, and 2 support feet
The versatile Optical bench kit is designed for comprehensive study of image formation, focal lengths, and image magnification. Featuring a 120 cm bench for precise lens alignment, the set includes essential components such as an LED letter F, object screen, two 200 mm focal length lenses (convex and concave), two 100 mm focal length lenses (concave and convex), and six mounts.
Ideal for educational settings, this kit facilitates hands-on exploration of optical principles with ease and clarity. Perfect for a Physical science classroom or homeschool. Educational use: The optical bench kit is ideal for science and physics classrooms, facilitating a hands-on approach to learning optics.
Comprehensive study: Designed for in-depth exploration of image formation, focal lengths, and magnification in optical systems.
Key components: Includes a 100 cm bench for precise lens alignment, LED letter F, object screen, two 200 mm focal length lenses (convex and concave), two 120 mm focal length lenses (concave and convex), and six mounts.
Physics and optics: Perfect for demonstrating key principles of optics and light behavior in educational settings.
Classroom ready: This versatile kit supports clear and easy exploration of optical principles, making it an essential tool for effective teaching.
 : This product can expose you to chemicals including nickel, lead, and styrene, which are known to the State of California to cause cancer, birth defects, or other reproductive harm. These products are not toys. Use in a laboratory or educational setting only. Not for children 13 and under.