ContactUsLinkComponent
NANOCOLOR® Photometric Tube Tests, Polyoxycarboxylic Acids
  77578-556
New Product
 :  
undefined
NANOCOLOR® Photometric Tube Tests, Polyoxycarboxylic Acids
  77578-556
 :  985070
New Product
 :  
  • Chemical method:
    Turbidimetric
  • Description:
    POC 200 (Polyoxycarboxylic acids)
  • Cat. no.:
    77578-556
  • Range:
    20 - 200 mg/L POC
  • No. of Tests:
    20
A pre-dosed NANOCOLOR® tube test for photometric determination of polyoxycarboxylic acids in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (OD) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.