NANOCOLOR® Photometric Tube Tests, Total Chromium
77578-588
New Product
:
- Chemical method:Diphenylcarbazide
- Description:Total Chromium 2
- Cat. no.:77578-588
- Range:0.05 - 2.00 mg/L Cr
- No. of Tests:20
A pre-dosed NANOCOLOR® tube test for photometric determination of chromium in water.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in 16 mm (OD) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.