NANOCOLOR® Photometric Tube Tests, Nickel
Catalog # 77578-518
Supplier: Macherey-Nagel
New Product
CAS Number:
Specifications
- Chemical method:Dimethylglyoxime
- Description:Nickel 4
- Cat. no.:77578-518
- Range:0.10 - 7.00 mg/L Ni²⁺
- No. of Tests:20
Specifications
About this item
A pre-dosed NANOCOLOR® tube test for photometric determination of nickel in water.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in 16 mm (ext.Ø) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.