Crystal Digital PCR® Assay for NRAS (drop-off G12-G13)
Supplier: STILLA TECHNOLOGIES, INC.
New Product
Certificates
About this item
Assay-specific crystal flex probes and primers for the detection of NRAS mutations and its positive control. The NRAS (drop-off G12-G13) Crystal Digital PCR® assay is a 10X assay designed to detect and quantify mutations in codons 12 and 13 of the NRAS gene using the ruby chip. NRAS is pivotal in regulating cell signaling pathways implicated in cancer development, notably melanoma and colorectal cancer.
- Ready-to-use assay reagents to detect multiple targets in a single reaction
- Crystal flex probe detection chemistry for flexible assay configuration
- Same set of crystal universal reporters to detect any target in different assays
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, Prism3, ruby chip
Channels: Blue, red
Required reagents: Crystal universal reporter 3 and naica® PCR MIX 10X
Targets: NRAS G12-G13
Caution: For research use only. Not for use in diagnostic procedures.
Related Information
Stilla Technologies Crystal Flex Probes and Crystal Universal Reporters
Product Literature
Frequently Bought Together