Orders LinkContactUsLinkComponent
Cornerstone Workbenches
Cornerstone Workbenches
Supplier:   Treston
Cornerstone Workbenches
Supplier:  Treston
Certificates
Cornerstone Workbenches
Cornerstone Workbenches
4 Options Available Below
Product Details & Documents
The American-made Treston® Cornerstone industrial workbench combines ergonomic design, heavy-duty steel construction, and unmatched customization options. With a 1500 lbs load capacity and a wide range of modular accessories, the Cornerstone is perfect for any workplace.
  • Fast, easy assembly
  • Modern design and ESD compatible
  • Four legged steel structure
  • Up to 1500 lbs. load capacity
  • Select frames QuickShip eligible
Adjustment range 29 to 35", in 1" increments (leg extenders also available for taller height adjustment).
Color: Light gray.
With 10 year warranty.
Ordering information: Installation is required. Assembly videos available.
Product Family Options
Product Information
  • W×D
    W×D×H
    Weight
    Availability
    Price
  • 77575-694
    Cornerstone frame
    Cornerstone frame
    96×24"
    1000 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,450.36
    $1,450.36
    Each
       $1,450.36
    Minimum order quantity is 1
    Supplier Number: CSB2496-49
  • 77575-702
    Cornerstone frame
    Cornerstone frame
    96×30"
    1000 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,480.76
    $1,480.76
    Each
       $1,480.76
    Minimum order quantity is 1
    Supplier Number: CSB3096-49
  • 77575-710
    Cornerstone frame
    Cornerstone frame
    96×36"
    1000 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,655.69
    $1,655.69
    Each
       $1,655.69
    Minimum order quantity is 1
    Supplier Number: CSB3696-49
  • 77575-712
    Cornerstone leg extender, 30 - 40"
    Cornerstone leg extender, 30 - 40"
    1.73×1.73×17.9"
    9 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $277.27
    $277.27
    Each
       $277.27
    Minimum order quantity is 1
    Supplier Number: CSBLE-49