ContactUsLinkComponent
ReadiLink™ Rapid iFluor® 488 Antibody Labelling Kit
  77462-908
 :  
undefined
ReadiLink™ Rapid iFluor® 488 Antibody Labelling Kit
  77462-908
 :  5702
 :  
  • Host:
  • Size:
    3×1 mg
  • Environmentally Preferable:
  • Cat. no.:
    77462-908
ReadiLink™ Rapid antibody labelling kits, designed for production scale, provide a convenient and efficient method for labelling large volumes of antibodies with our superior iFluor® dyes.
  • Robust protocol
  • Consistent results
  • Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
 : Shipped at RT.
 : Research use only.