ContactUsLinkComponent
Buccutite™ Rapid PerCP Antibody Labelling Kit
Buccutite™ Rapid PerCP Antibody Labelling Kit
Catalog # 77462-880
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
Buccutite™ Rapid PerCP Antibody Labelling Kit
Catalog # 77462-880
Supplier:  AAT BIOQUEST INC
Supplier Number:  5407
CAS Number:  
Specifications
  • Size:
    2
  • Cat. no.:
    77462-880
  • Includes:
    2 labellings
Specifications
About this item
Production scale optimized for labelling 1 mg antibody per reaction. Buccutite™ Rapid PerCP antibody labelling kits, designed for large-scale production, provide a streamlined approach for labelling antibodies with PerCP, APC, PE and iFluor® tandem dyes.
  • Robust protocol
  • Consistent results
  • Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Delivery: Shipped with blue ice.
Caution: Research use only.