ContactUsLinkComponent
Buccutite™ Rapid trFluor™ D2 Acceptor Antibody Labelling Kit
Buccutite™ Rapid trFluor™ D2 Acceptor Antibody Labelling Kit
Catalog # 77461-428
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
Buccutite™ Rapid trFluor™ D2 Acceptor Antibody Labelling Kit
Catalog # 77461-428
Supplier:  AAT BIOQUEST INC
Supplier Number:  1302
CAS Number:  
Specifications
  • Size:
    2
  • Cat. no.:
    77461-428
  • Includes:
    2 labellings
Specifications
About this item
Microscale optimized for labelling 100 µg antibody per reaction. Many biological compounds present in cells, serum, or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
  • Robust protocol
  • Consistent results
  • 12-month shelf life
Designed to provide high quality labelled antibodies for multiple downstream applications.
Delivery: Shipped at RT.
Caution: Research use only.