ContactUsLinkComponent
Cell Meter™ Glucose Uptake Imaging Kit
Cell Meter™ Glucose Uptake Imaging Kit
Catalog # 77461-928
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
Cell Meter™ Glucose Uptake Imaging Kit
Catalog # 77461-928
Supplier:  AAT BIOQUEST INC
Supplier Number:  23501
CAS Number:  
Specifications
  • Size:
    100 Tests
  • Cat. no.:
    77461-928
  • No. of Tests:
    100 tests
Specifications
About this item
Red Fluorescence. The Cell Meter™ glucose Uptake imaging kit provides a simple and direct method for quantifying glucose uptake across diverse cellular contexts.
  • Ex/Em = 651/670 nm
  • Quantification via both fluorescence microscopy and flow cytometry techniques
  • Six-month shelf life
This kit employs Glutite™ Red 670, a cell-permeable fluorescent glucose tracer, enabling precise molecular sensing and bioimaging via GLUT transporters.
Delivery: Shipped at RT.
Caution: Research use only.