Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (APC (Allophycocyanin)/iFluor® 750)
Supplier: AAT BIOQUEST INC
Certificates
About this item
Anti-human secondary antibodies have well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.
- Affinity purified
- Easily reconstituted
Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence).
Caution: For research use only.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)/iFluor® 750
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Human
Specifications
- Catalog No:
- 77462-714
- 77462-712
- Antigen symbol:
- IgG
- IgG
- Antigen name:
- Immunoglobulin G
- Immunoglobulin G
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- APC (Allophycocyanin)/iFluor® 750
- APC (Allophycocyanin)/iFluor® 750
- Reactivity:
- Human
- Human
- Host:
- Goat
- Goat
- Isotype:
- IgG (H, L)
- IgG (H, L)
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Cross adsorption:
- Minimal cross reaction to human, horse, mouse, rabbit and bovine
- Minimal cross reaction to human, horse, mouse, rabbit and bovine
- Storage temperature:
- Freeze (<–15 °C); minimize light exposure
- Freeze (<–15 °C); minimize light exposure
- Shipping temperature:
- 20
- 20
Specifications
Frequently Bought Together