Order Entry
Canada
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (iFluor® 594)
Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (iFluor® 594)
Supplier:  AAT BIOQUEST INC
Certificates
undefined
Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (iFluor® 594)
Supplier:  AAT BIOQUEST INC

Specifications

  • Catalog No:
  • 77462-480
  • 77462-482
  • Antigen symbol:
  • IgG
  • IgG
  • Antigen name:
  • Immunoglobulin G
  • Immunoglobulin G
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • iFluor® 594
  • iFluor® 594
  • Reactivity:
  • Human
  • Human
  • Host:
  • Goat
  • Goat
  • Isotype:
  • IgG (H, L)
  • IgG (H, L)
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • Minimal cross reaction to human, horse, mouse, rabbit and bovine
  • Minimal cross reaction to human, horse, mouse, rabbit and bovine
  • Storage temperature:
  • Freeze (<–15 °C); minimize light exposure
  • Freeze (<–15 °C); minimize light exposure
  • Shipping temperature:
  • 20
  • 20

Specifications

Frequently Bought Together