Cleanroom Arc Flash Coveralls, 8 Cal
Supplier: Sellstrom
New Product
Certificates
About this item
Clean room arc rated coveralls are designed to meet specific cleanliness requirements for use in clean room environments.
- Coveralls are particle-free
- Made from non-shedding materials (flame-resistant Nomex fabric) to prevent contamination of the environment
- Provide protection against arc flash hazards and electrostatic discharge
- Generous fit for maximum comfort while performing electrical work tasks
- Dimension: 21×5×13"
The coveralls are designed to be comfortable and breathable while providing the necessary protection and cleanliness required in these sensitive environments. These garments may be worn by workers in industries such as semiconductor manufacturing, pharmaceuticals, and electronics assembly, among others.
Certifications: Meets ANSI/ISEA 125 Level 2 Conformity and Arc Flash PPE Category 2 standards with an arc rating of 8 cal/cm². Please refer to NFPA 70E or CSA Z462 Standards for specific selection requirements, ASTM F1506, ASTM F1959.
Specifications
- Catalog No:
- 77575-478
- 77575-480
- 77575-494
- 77575-536
- 77575-516
- 77575-488
- 77575-504
- 77575-530
- Model:
- CR-CVL
- CR-CVL
- CR-CVL
- CR-CVL
- CR-CVL
- CR-CVL
- CR-CVL
- CR-CVL
- Garment application:
- Industrial, Electronics/semi-conductor
- Industrial, Electronics/semi-conductor
- Industrial, Electronics/semi-conductor
- Industrial, Electronics/semi-conductor
- Industrial, Electronics/semi-conductor
- Industrial, Electronics/semi-conductor
- Industrial, Electronics/semi-conductor
- Industrial, Electronics/semi-conductor
- Risk area:
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Fabric:
- Nomex
- Nomex
- Nomex
- Nomex
- Nomex
- Nomex
- Nomex
- Nomex
- Weight:
- 3.08 lbs.
- 3.08 lbs.
- 3.08 lbs.
- 3.08 lbs.
- 3.08 lbs.
- 3.08 lbs.
- 3.08 lbs.
- 3.08 lbs.
Specifications
Frequently Bought Together