ContactUsLinkComponent
Countstar Castor High-Throughput Cell Analyzers
Countstar Castor High-Throughput Cell Analyzers
Supplier:  ALIT LIFETECH INC
New Product
Certificates
undefined
Countstar Castor High-Throughput Cell Analyzers
Supplier:  ALIT LIFETECH INC
New Product
Specifications
  • Catalog No:
  • 77591-746
  • 77591-748
  • 77591-750
  • Wavelength:
  • Ex: 395/15 nm, Em: 460/20 nm;
    Ex: 480/30 nm, Em: 535/40 nm;
    Ex: 545/15 nm, Em: 590/25 nm;
    (fluorescent channels)
  • Ex: 395/15 nm, Em: 460/20 nm;
    Ex: 480/30 nm, Em: 535/40 nm;
    Ex: 545/15 nm, Em: 590/25 nm;
    (fluorescent channels)
  • Ex: 395/15 nm, Em: 460/20 nm;
    Ex: 480/30 nm, Em: 535/40 nm;
    Ex: 545/15 nm, Em: 590/25 nm;
    (fluorescent channels)
  • Magnification:
  • 4×, 10×
  • 4×, 10×
  • 4×, 10×
  • Camera:
  • 10 MP, CMOS
  • 10 MP, CMOS
  • 10 MP, CMOS
  • W×D×H:
  • 21.8×21.3×19"
  • 21.8×21.3×19"
  • 21.8×21.3×19"
  • Weight:
  • 80.4 lbs.
  • 80.4 lbs.
  • 80.4 lbs.