Orders LinkContactUsLinkComponent
Lysis Buffer, FLEXLyse
Lysis Buffer, FLEXLyse
FlexLyse, 100 ml, 1 Vial
Catalog # CA77575-364
Supplier:   Beckman Coulter
Supplier #:  C54313
Lysis Buffer, FLEXLyse
Catalog # CA77575-364
Supplier:  Beckman Coulter
CAS Number:  
Lysis Buffer, FLEXLyse
FlexLyse, 100 ml, 1 Vial
CAS Number:  
Price
$1,550.39
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Whole blood assays are our best approximation of physiological conditions in research flow cytometry. Density gradient leukocyte isolation or bulk lysing procedures are common to remove red blood cells, but decompose the native material prior to antigen detection. Common lysing buffers avoid this decomposition and eliminate the gradient-related preanalytical workload, but also increase sample volume beyond the single-well capacity of many plate formats, limiting throughput and automation options.
  • FLEXLyse processes blood samples at much smaller total volumes than common lysing buffers, enabling automation of whole blood lysis procedures:
  • For 96 shallow well plates with 25 to 40 μl of whole blood/well (2000 tests/unit FLEXLyse)
  • For 96 deep well plates with 50 to 100 μl of whole blood/well (330 tests/unit FLEXLyse)
  • Leave prepared samples for up to 48 hrs at room temperature on liquid handler deck*
  • Avoid integration of centrifuges with no-wash protocols*
  • Combine FLEXLyse with dry DURA innovations reagents for streamlined automated protocols - no need for reagent cooling
  • Liquid format
*No-wash protocol established with antibody combination according to DURAClone IM phenotyping basic, stained in EDTA-anticoagulated whole blood. For different materials and antibody combinations, users are responsible for verifying prepared sample stability and no-wash compatibility.
Specifications
Specifications
  • Pack type:
    1 Vial
  • Description:
    FlexLyse
  • Size:
    100 ml
Specifications