Ward's® Caliche
About this item
Hardened natural cement of calcium carbonate that binds other material such as gravel, sand, clay, and silt.
Collected in Arizona
- Multiple sizes and pack options to fit your specific needs
- Cut by Ward's geologists to guarantee top market quality
- Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
All specimen dimensions are considered an average size range.
2 Options Available Below
Product Details & Documents
Hardened natural cement of calcium carbonate that binds other material such as gravel, sand, clay, and silt.
Collected in Arizona
- Multiple sizes and pack options to fit your specific needs
- Cut by Ward's geologists to guarantee top market quality
- Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Product Information
- Specimen dimensionsAvailabilityPrice
- 470323-126Caliche, bulk pack, 1 kg
- 470323-116Caliche, student specimens
Storage
Product Information
- Catalog #DescriptionAvailabilityPrice