Orders LinkContactUsLinkComponent
Klinger Multiple Ray Projector (Suction)
Klinger Multiple Ray Projector (Suction)
Klinger multiple ray projector (suction)
Catalog # 470336-010
Supplier:   Klinger
Supplier #:  KO4125
Klinger Multiple Ray Projector (Suction)
Catalog # 470336-010
Supplier:  Klinger
Klinger Multiple Ray Projector (Suction)
Klinger multiple ray projector (suction)
Price
$2,059.29
Log in for availability
Each
Product Details & Documents
Use this ray box on a whiteboard to vividly demonstrate optical principles.
  • Produce rays using an incandescent lamp rather than LED or laser
  • Attach ray projector to veritcal surface using suction cups
  • Great for use on non-magnetic surfaces
This projector provides up to five well collimated light beams that trace clear optical paths along a chalkboard. The fixed central beam establishes the optical axis. The other rays may be adjusted to desired angles to demonstrate spherical aberration.
Delivery information: Includes lamp (12 V, 55 W).
Specifications
  • Description:
    Klinger multiple ray projector (suction)