Klinger Laser Boxes
About this item
Perform many different lessons around reflection, refraction, diffraction, and interference.
- Class 3R lasers
- Line projection lens installed
- Wavelengths: 635 nm (red), 532 nm (green), and 450 nm (blue)
- 2× AA batteries included
Klinger lasers are classroom safe and are designed for teaching and learning about light, lasers, and optics. These lasers can be used just like a laser pointer or on a white board with their powerful magnets to keep them in place. Place multiple lasers together side by side to create a Ray Box for even more versatility and flexibility.
7 Options Available Below
Product Details & Documents
Perform many different lessons around reflection, refraction, diffraction, and interference.
- Class 3R lasers
- Line projection lens installed
- Wavelengths: 635 nm (red), 532 nm (green), and 450 nm (blue)
- 2× AA batteries included
Klinger lasers are classroom safe and are designed for teaching and learning about light, lasers, and optics. These lasers can be used just like a laser pointer or on a white board with their powerful magnets to keep them in place. Place multiple lasers together side by side to create a Ray Box for even more versatility and flexibility.
Product Family Options
Product Information
- AvailabilityPrice
- 470335-996Klinger laser box, green, singleSupplier Number: LB-GR-KL
- 470335-998Klinger laser box, red, singleSupplier Number: LB-RD-KL
- 470336-000Klinger laser box, blue, singleSupplier Number: LB-BL-KL
- 470335-988Klinger laser boxes, green, set of 3Supplier Number: 3G-LB-KL
- 470335-990Klinger laser boxes, red, set of 3Supplier Number: 3R-LB-KL
- 470335-992Klinger laser boxes, blue, set of 3Supplier Number: 3B-LB-KL
- 470335-994Klinger laser boxes, multi-color, set of 3Supplier Number: MLTI-LB-KL
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.