Gelite™ 100 bp DNA Ladder
77461-672
:
- Description:Gelite™ 100 bp DNA ladder
- Size:0.5 ml
- Cat. no.:77461-672
The Gelite™ 100 bp DNA Ladder is designed for sizing and estimating the mass of double-stranded DNA and PCR products within the 100 to 1500 bp range on agarose or polyacrylamide gels.
- Stable for 12 months
Quality tested in-house.
: For research use only.