FluoroQuest™ Antifade Mounting Medium
Catalog # 77461-728
Supplier: AAT BIOQUEST INC
CAS Number:
Specifications
- Description:FluoroQuest™ Antifade mounting medium
- Size:5 ml
- Cat. no.:77461-728
Specifications
About this item
FluoroQuest™ Antifade mounting medium is a specialized and stable formula that effectively preserves fluorescence by preventing rapid photobleaching of fluorescent proteins and dyes.
- Stable for 12 months
Quality tested in-house.
Caution: For research use only.