Recombinant Lysostaphin (from E. coli)
Catalog # 77540-186
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Size:10 mg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:Glycyl-glycine endopeptidase
- Protein/peptide name:Lysostaphin
- Purity:96.9% as determined by RP-HPLC
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77540-186