Human Recombinant LGR6 (from E. coli)
77540-132
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:50 µg
- Storage conditions:4 °C
- Reconstitution instructions:Please reconstitute protein in deionized sterile water to a concentration of 1.0 mg/ml. We recommend to add 5 - 50% of glycerol (final concentration) and aliquot for long-term storage at −20/−80 °C.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:leucine-rich repeat containing G protein-coupled receptor 6
- UniProtKB:Q9HBX8
- Protein/peptide name:LGR6
- Purity:Greater than 85% as determined by SDS-PAGE
- Molecular weight:61.6 kDa
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77540-132