Human Recombinant KPNA2 (from E. coli)
Catalog # 77540-062
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:50 µg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:karyopherin alpha 2 (RAG cohort 1, importin alpha 1)
- UniProtKB:P52292
- Protein/peptide name:KPNA2
- Purity:>90% by SDS-PAGE
- Endotoxin level:Low
- Concentration:1 mg/ml(determined by Bradford assay)
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77540-062