Human Recombinant IFNGR1 (from HEK293 cells)
77539-850
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:25 µg
- Storage conditions:4 °C
- Reconstitution instructions:It is recommended that 0.5 ml of sterile phosphate-buffered saline be added to the vial.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:IFNGR1 interferon gamma receptor 1
- UniProtKB:P15260
- Protein/peptide name:IFNGR1
- Purity:>95% as determined by SDS-PAGE and visualized by Coomassie brilliant blue
- Molecular weight:IFN-gamma R1 - Fc Chimera migrates as a broad band between 60 and 85 kDa on SDS-PAGE due to post-translation modifications, in particular glycosylation.
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-850