Human Recombinant HMGB1 (from CHO cells)
Catalog # 77539-770
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Size:50 µg
- Storage conditions:4 °C
- Endotoxin content:<0.06 EU/µg as determined by lal test
- Biological activity:Measured in a competitive binding assay
- Reconstitution instructions:Reconstitute at 100 ug/ml in sterile PBS
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:high mobility group box 1
- UniProtKB:P09429
- Protein/peptide name:HMGB1
- Purity:≥98% by SDS-PAGE under reducing conditions
- Molecular weight:48 KDa
- Sequence:Arg24-Glu215
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-770