Human Recombinant HIP11 (from E. coli)
Supplier: Creative Biomart
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:Proinsulin HIP 11
- Protein/peptide name:HIP11
- Purity:>90% as determined by SDS-PAGE
- Molecular weight:~13.35 kDa
- Sequence:MFVNQHLCGSHLVEALYLVCGERGFFYTPKTRREAEDLQVGQVELGGGPGAGSLQPLALEAEDLQVGQVELGGGPGAGSLQPLALEGSLQEGSLQKRGIVEQCCTSICSLYQLENYCNHHHHHH
- Endotoxin level:Low
- Concentration:0.3 µg/µl
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
Specifications
Frequently Bought Together