Order Entry
Canada
ContactUsLinkComponent
Human Recombinant HAVCR2 (from HEK293 cells)
Human Recombinant HAVCR2 (from HEK293 cells)
Catalog # 77539-684
Supplier:  Creative Biomart
CAS Number:  
undefined
Human Recombinant HAVCR2 (from HEK293 cells)
Catalog # 77539-684
Supplier:  Creative Biomart
Supplier Number:  HAVCR2-3166H
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    100 µg
  • Storage conditions:
    4 °C
  • Endotoxin content:
    <1.0 EU/µg protein as determined by the LAL method
  • Biological activity:
    Captured Anti-TIM-3 Mab (Human IgG4) on proA Chip can bind TIM-3 with an affinity constant of 2.351 nM as determined in a SPR assay (Biacore T200)
  • Reconstitution instructions:
    It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
  • Endotoxin-free:
    No
  • Carrier-free:
    No
  • Protease-free:
    No
  • Animal-free:
    No
  • Protein synonyms:
    hepatitis A virus cellular receptor 2
  • UniProtKB:
    Q8TDQ0
  • Protein/peptide name:
    HAVCR2
  • Purity:
    >95% as determined by SDS-PAGE
  • Molecular weight:
    The recombinant human HAVCR2 consists of 190 amino acids and predicts a molecular mass of 21.3 kDa. In SDS-PAGE under reducing conditions, the apparent molecular mass of human HAVCR2 is approximately 42.8 kDa due to glycosylation.
  • Endotoxin level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping temperature:
    20
  • Cat. no.:
    77539-684

Specifications

About this item