Recombinant GFP (from E. coli)
Catalog # 77539-596
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Size:25 µg
- Storage conditions:4 °C
- Endotoxin content:<1.0 EU/µg (determined by the LAL method)
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- UniProtKB:P42212
- Protein/peptide name:GFP
- Purity:>95% by SDS-PAGE
- Sequence:SMSKGEELFTGVVPILVELDGDVNGHKFSVSGEGEGDATYGKLTLKFICTTGKLPVPWPTLVTTFSYGVQCFSRYPDHMKQHDFFKSAMPEGYVQERTIFFKDDGNYKTRAEVKFEGDTLVNRIELKGIDFKEDGNILGHKLEYNYNSHNVYIMADKQKNGIKVNFKIRHNIEDGSVQLADHYQQNTPIGDGPVLLPDNHYLSTQSALSKDPNEKRDHMVLLEFVTAAGITHGMDELYKTRTRPLEQKLISEEDLAANDILDYKDDDDKV
- Endotoxin level:Low
- Concentration:0.4 mg/ml
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-596