Mouse Recombinant Fgf21 (from HEK293 cells)
Catalog # 77539-476
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:20 µg
- Storage conditions:4 °C
- Endotoxin content:<1.0 EU/µg of the protein as determined by the LAL method
- Reconstitution instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:fibroblast growth factor 21
- Protein/peptide name:Fgf21
- Purity:>85% as determined by SDS-PAGE
- Molecular weight:The secreted recombinant mouse FGF21 consists of 193 amino acids and has a calculated molecular mass of 21.3 kDa. As a result of glycosylation, the apparent molecular mass of rm FGF21 is approximately 25 kDa in SDS-PAGE under reducing conditions.
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-476