Human Recombinant FCER1A (from HEK293 cells)
Catalog # 77539-448
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:10 µg
- Storage conditions:4 °C
- Endotoxin content:<1.0 EU/µg by the LAL method
- Biological activity:Immobilized Immunoglobulin E, Human Plasma at 5 μg/ml (100 μl/well) can bind Human Fc epsilon RI alpha, His Tag with a linear range of 0.4 - 3 ng/ml.
- Reconstitution instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:receptor for alpha polypeptide|high affinity I|Fc fragment of IgE
- UniProtKB:P12319
- Protein/peptide name:FCER1A
- Purity:>95% as determined by SDS-PAGE. >90% as determined by SEC-MALS.
- Molecular weight:The protein has a calculated MW of 22.9 kDa. The protein migrates as 37 - 60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-448