Human Recombinant EGFR (from HEK293 cells)
Catalog # 77539-348
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:50 µg
- Storage conditions:4 °C
- Endotoxin content:<1.0 EU/µg by the LAL method
- Biological activity:Immobilized Human EGF, Tag Free at 10 μg/ml (100 μl/well) can bind Recombinant Human EGFR Protein, Fc-tagged with a linear range of 0.2 - 3.2 μg/ml (QC tested). Recombinant Human EGFR Protein, Fc-tagged at 1 μg/ml (100 μl/well) can bind Cetuximab with a linear range of 0.1 - 2 ng/ml (Routinely tested).
- Reconstitution instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 mg/ml. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:epidermal growth factor receptor
- UniProtKB:P00533
- Protein/peptide name:EGFR
- Purity:>95% as determined by SDS-PAGE
- Molecular weight:95 kDa
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-348