Human Recombinant EGF (from E. coli)
77539-340
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:1 mg
- Storage conditions:4 °C
- Endotoxin content:<0.3 ng/µg (0.3 IEU/µg) determined by LAL test
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:Epidermal growth factor
- UniProtKB:P01133
- Protein/peptide name:EGF
- Purity:>95% as determined by SDS-PAGE and SEC-HPLC
- Molecular weight:6200 Da
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-340