Human Recombinant DLL1 (from CHO cells)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Storage conditions:4 °C
- Biological activity:Inhibits adipogenesis of 3T3L-1 cells. Induces Hes-1 in 3T3L-1 cells.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:delta-like 1 (Drosophila)
- UniProtKB:O00548
- Protein/peptide name:DLL1
- Purity:≥90% (SDS-PAGE)
- Molecular weight:~100 kDa (SDS-PAGE)
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
Frequently Bought Together