Cynomolgus Monkey Recombinant CXCL16 (from E. coli)
Catalog # 77539-244
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Cynomolgus monkey
- Size:100 µg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:chemokine (C-X-C motif) ligand 16
- Protein/peptide name:CXCL16
- Purity:>90%, by SDS-PAGE
- Molecular weight:~55.5 KDa, reducing conditions
- Sequence:MNWSHPQFEKSSGSSGGHHHHHHGGSGGSGSDSEVNQEAKPEVKPEVKPETHINLKVSDGSSEIFFKIKKTTPLRRLMEAFAKRQGKEMDSLRFLYDGIRIQADQAPEDLDMEDNDIIEAHREQIGGMGVGGRDVRAAGARAPPPPRPRPRRGLTDFPAVLTRHVTQCRRPAGKRKRPGRWWRRRAGGAVAAGGPPDLGRLGGHPRAGPSSCCLCALLPGRSLGAGARGTGARGRGAPPPGLSPRRWRWGAAAARPAVPTALSVRKVRAREEGASSSRRTPHPRTPTAAAARRPGGPGREQLDAALRSGLAPDMGAGPLVLLAALALLTRPGDGNEGSVTGSCYCDKAISSGSPPTAELMAHLRKHLRVYQRCNSYVRFQLRMRSVCGGSKDPWVQQLMSCLDRKECGSADSRSVAPQEQLPPLSTQVPEPTERAPLDMGSPAQTYLPPAWRSTYLPTALQSTRQPVFPAGTLSLEKKLTHTSEIATSTVGHSLRTGSEAGESQKQQIKRVEPTAGTS
- Endotoxin level:Low
- Concentration:0.1 mg/ml
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-244