Ginkgo biloba Native EP7 (from Ginkgo Biloba L. Leaf)
77539-136
:
- Pack type:Pack
- Conjugation:Unconjugated
- Protein/peptide type:Native
- Source:Leaf
- Species:Ginkgo biloba
- Size:1 kg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:EP7
- Protein/peptide name:EP7
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-136