Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant CD79B (from HEK293 cells)
 
undefined
Human Recombinant CD79B (from HEK293 cells)

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    4 °C
  • Endotoxin content:
    <1.0 EU/µg by the LAL method
  • Reconstitution instructions:
    It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
  • Endotoxin-free:
    No
  • Carrier-free:
    No
  • Protease-free:
    No
  • Animal-free:
    No
  • Protein synonyms:
    CD79b molecule, immunoglobulin-associated beta
  • UniProtKB:
    P40259
  • Protein/peptide name:
    CD79B
  • Purity:
    >92% as determined by SDS-PAGE
  • Molecular weight:
    The protein has a calculated MW of 41.8 kDa. The protein migrates as 50 - 65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
  • Endotoxin level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping temperature:
    20

 

Frequently Bought Together