Human Recombinant CD22 (from HEK293 cells)
77538-904
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:5 µg
- Storage conditions:4 °C
- Endotoxin content:<1.0 EU/µg by the LAL method
- Biological activity:Immobilized Human Siglec-2, Fc Tag at 2 μg/ml (100 μl/well) can bind Monoclonal Anti-Human Siglec-2 Antibody, Human IgG1 with a linear range of 2 - 62.5 ng/ml.
- Reconstitution instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:CD22 molecule
- UniProtKB:P20273
- Protein/peptide name:CD22
- Purity:>95% as determined by SDS-PAGE
- Molecular weight:This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 101.6 kDa. The protein migrates as 125 - 150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77538-904