Order Entry
Canada
ContactUsLinkComponent
Human Recombinant CD19 (from HEK293 cells)
Human Recombinant CD19 (from HEK293 cells)
Supplier:  Creative Biomart
Certificates
undefined
Human Recombinant CD19 (from HEK293 cells)
Supplier:  Creative Biomart

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    4 °C
  • Endotoxin content:
    <1.0 EU/µg by the LAL method
  • Biological activity:
    Immobilized Monoclonal Anti-Human CD19 Antibody, Mouse IgG2a at 2 μg/ml (100 μl/well) can bind Human CD19 (20-291), His Tag with a linear range of 10 - 78 ng/ml. Immobilized Human CD19 (20-291), His Tag at 2 μg/ml (100 μl/well) can bind different Anti-CD19 antibodies with high affinity.
  • Reconstitution instructions:
    It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
  • Endotoxin-free:
    No
  • Carrier-free:
    No
  • Protease-free:
    No
  • Animal-free:
    No
  • Protein synonyms:
    CD19 molecule
  • UniProtKB:
    P15391
  • Protein/peptide name:
    CD19
  • Purity:
    >90% as determined by SDS-PAGE
  • Molecular weight:
    This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 32.0 kDa. The protein migrates as 47 - 58 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
  • Endotoxin level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping temperature:
    20

Specifications

Frequently Bought Together