Rat Recombinant Capn2 (from E. coli)
Catalog # 77538-830
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Size:50 µg
- Storage conditions:4 °C
- Biological activity:1.47 U/mg protein. One unit is defined as the amount of enzyme that cleaves 1 pmol of the caspase substrate N-succinyl-Leu-Tyr-7-amido-4-methylcoumarin per 15 min at 37 centigrade.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:calpain 2, (m/II) large subunit
- UniProtKB:Q07009
- Protein/peptide name:Capn2
- Purity:>90% by SDS-PAGE
- Sequence:MAGIAMKLAKDREAAEGLGSHERAIKYLNQDYETLRNECLEAGALFQDPSFPALPSSLGFKELGPYSSKTRGIEWKRPTEICADPQFIIGGATRTDICQGALGDCWLLAAIASLTLNEEILARVVPLDQSFQENYAGIFHFQFWQYGEWVEVVVDDRLPTKDGELLFVHSAEGSEFWSALLEKAYAKINGCYEALSGGATTEGFEDFTGGIAEWYELRKPPPNLFKIIQKALEKGSLLGCSIDITSAADSEAVTYQKLVKGHAYSVTGAEEVESSGSLQKLIRIRNPWGQVEWTGKWNDNCPSWNTVDPEVRANLTERQEDGEFWMSFSDFLRHYSRLEICNLTPDTLTCDSYKKWKLTKMDGNWRRGSTAGGCRNYPNTFWMNPQYLIKLEEEDEDDEDGERGCTFLVGLIQKHRRRQRKMGEDMHTIGFGIYEVPEELTGQTNIHLSKNFFLTTRARERSDTFINLREVLNRFKLPPGEYVLVPSTFEPHKNGDFCIRVFSEKKADYQTVDDEIEANIEEIEANEEDIGDGFRRLFAQLAGEDAEISAFELQTILRRVLAKREDIKSDGFSIETCKIMVDMLDEDGSGKLGLKEFYILWTKIQKYQKIYREIDVDRSGTMNSYEMRKALEEAGFKLPCQLHQVIVARFADDELIIDFDNFVRCLVRLEILFKIFKQLDPENTGTIQLDLISWLSFSVLHHHHHHMHHHHHHHYSNIEANESEEERQFRKLFVQLAGDDMEVSATELMNILNKVVTRHPDLKTDGFGIDTCRSMVAVMDSDTTGKLGFEEFKYLWNNIKKWQGIYKRFDTDRSGTIGSNELPGAFEAAGFHLNQHIYSMIIRRYSDETGNMDFDNFISCLVRLDAMFRAFRSLDKNGTGQIQVNIQEWLQLTMYS
- Endotoxin level:Low
- Concentration:1.2 mg/ml by Bradford
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77538-830