Human Native C4BPB (from Plasma)
Catalog # 77538-802
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Native
- Source:Plasma
- Species:Human
- Size:250 µg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:C4b-Binding Protein Beta Chain
- UniProtKB:P20851
- Protein/peptide name:C4BPB
- Purity:≥95% by SDS-PAGE
- Molecular weight:~570000 (Whole molecule), ~70 - 75000 (Alpha chains), ~40 - 45000 (Beta chain)
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77538-802